Assay Details
Gene Symbol: SURF1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_021071112.1 | XP_020926771.1 | 356 | 103 | ||
XM_021071119.1 | XP_020926778.1 | 359 | 103 | |||
XM_021071124.1 | XP_020926783.1 | 601 | 103 | |||
GenBank mRNA | AK391958.1 | - | 4 - 5 | 354 | 103 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
