Assay Details
Gene Symbol: SURF1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_021071112.1 | XP_020926771.1 | 872 | 79 | ||
XM_021071119.1 | XP_020926778.1 | 875 | 79 | |||
XM_021071124.1 | XP_020926783.1 | 1117 | 79 | |||
GenBank mRNA | AK391958.1 | - | 8 - 9 | 870 | 79 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Panther Classification:
