Assay Details
Gene Symbol: PRSS33 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_021086741.1 | XP_020942400.1 | 1216 | 65 | ||
GenBank mRNA | AK392255.1 | - | 4 - 5 | 652 | 65 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Panther Classification:
