Assay Details
Gene Symbol: GTF3C6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003121346.4 | XP_003121394.2 | 253 | 120 | ||
XM_005652686.3 | XP_005652743.1 | 208 | 120 | |||
GenBank mRNA | AK392298.1 | - | 2 - 3 | 158 | 120 |

More Information
Set Membership: |
![]() ![]() ![]() |
