Assay Details
Gene Symbol: TEDC2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003124741.5 | XP_003124789.2 | 733 | 81 | ||
XM_005655185.3 | XP_005655242.1 | 733 | 81 | |||
XM_005655186.3 | XP_005655243.1 | 733 | 81 | |||
GenBank mRNA | AK392324.1 | - | 5 - 6 | 706 | 81 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
