Assay Details
Gene Symbol: TEDC2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005655186.3 | XP_005655243.1 | 1030 | 114 | ||
GenBank mRNA | AK392324.1 | - | 8 - 9 | 1003 | 114 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
