Assay Details
Gene Symbol: TEDC2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003124741.5 | XP_003124789.2 | 1217 | 84 | ||
XM_005655185.3 | XP_005655242.1 | 1307 | 84 | |||
XM_005655186.3 | XP_005655243.1 | 1214 | 84 | |||
XM_013987845.2 | XP_013843299.1 | 1253 | 84 | |||
XM_021086763.1 | XP_020942422.1 | 1163 | 84 | |||
GenBank mRNA | AK392324.1 | - | 9 - 10 | 1187 | 84 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
