Assay Details
Gene Symbol: COPRS | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003131747.4 | XP_003131795.1 | 214 | 97 | ||
GenBank mRNA | AK392391.1 | - | 1 - 2 | 170 | 97 | |
AY610442.1 | - | 1 - 2 | 117 | 97 |

More Information
Set Membership: |
![]() ![]() ![]() |
