Assay Details
Gene Symbol: COPRS | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003131747.4 | XP_003131795.1 | 278 | 69 | ||
GenBank mRNA | AK392391.1 | - | 2 - 3 | 234 | 69 |

More Information
Set Membership: |
![]() ![]() ![]() |
