Assay Details
Gene Symbol: CENPV | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_021066286.1 | XP_020921945.1 | 405 | 84 | ||
XM_021066287.1 | XP_020921946.1 | 405 | 84 | |||
GenBank mRNA | AK392474.1 | - | 1 - 2 | 75 | 84 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
