Assay Details
Gene Symbol: GDF10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003133105.4 | XP_003133153.1 | 592 | 79 | ||
GenBank mRNA | AK392607.1 | - | 1 - 2 | 573 | 79 |

More Information
Set Membership: |
![]() ![]() ![]() |
Panther Classification:
