Assay Details
Gene Symbol: BANK1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_021101719.1 | XP_020957378.1 | 285 | 90 | ||
GenBank mRNA | AK393699.1 | - | 1 - 2 | 182 | 91 |

More Information
Additional Information:
This assay is in the 5'UTR region of the XM_021101719.1 transcripts.
Set Membership: |
![]() ![]() ![]() |
