Assay Details
Gene Symbol: BANK1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_021101719.1 | XP_020957378.1 | 684 | 100 | ||
GenBank mRNA | AK393699.1 | - | 2 - 3 | 581 | 100 |

More Information
Set Membership: |
![]() ![]() ![]() |
