Assay Details
Gene Symbol: GLIS3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_021074379.1 | XP_020930038.1 | 2807 | 71 | ||
XM_021074388.1 | XP_020930047.1 | 2548 | 71 | |||
XM_021074390.1 | XP_020930049.1 | 2875 | 71 | |||
XM_021074393.1 | XP_020930052.1 | 2497 | 71 | |||
XM_021074396.1 | XP_020930055.1 | 2287 | 71 | |||
XM_021074403.1 | XP_020930062.1 | 2758 | 71 | |||
GenBank mRNA | AK393763.1 | - | 1 - 2 | 198 | 71 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
