Assay Details
Gene Symbol: POLE4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_021087319.1 | XP_020942978.1 | 271 | 78 | ||
XM_021087320.1 | XP_020942979.1 | 269 | 78 | |||
GenBank mRNA | AK393901.1 | - | 1 - 2 | 249 | 78 |

More Information
Set Membership: |
![]() ![]() ![]() |
Panther Classification:
