Assay Details
Gene Symbol: ALMS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005662464.3 | XP_005662521.1 | 12660 | 68 | ||
XM_005662465.3 | XP_005662522.1 | 10672 | 68 | |||
XM_013996028.2 | XP_013851482.1 | 12534 | 68 | |||
XM_021087373.1 | XP_020943032.1 | 12660 | 68 | |||
XM_021087374.1 | XP_020943033.1 | 9952 | 68 | |||
GenBank mRNA | AK396156.1 | - | 4 - 5 | 862 | 68 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
