Assay Details
Gene Symbol: PGBD1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003482141.4 | XP_003482189.1 | 1390 | 103 | ||
XM_021099793.1 | XP_020955452.1 | 1404 | 103 | |||
GenBank mRNA | AK396441.1 | - | 4 - 5 | 1121 | 103 |

More Information
Set Membership: |
![]() ![]() ![]() |
