Assay Details
Gene Symbol: TMEM79 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001927978.5 | XP_001928013.1 | 1134 | 85 | ||
GenBank mRNA | AK396516.1 | - | 2 - 3 | 896 | 85 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
