Assay Details
Gene Symbol: PTOV1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003127351.5 | XP_003127399.2 | 596 | 80 | ||
XM_021094765.1 | XP_020950424.1 | 597 | 80 | |||
GenBank mRNA | AK398188.1 | - | 2 - 3 | 133 | 80 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Panther Classification:
