Assay Details
Gene Symbol: PCBD2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_021084778.1 | XP_020940437.1 | 132 | |||
GenBank mRNA | AY610216.1 | - | 1 - 2 | 99 | 132 |

More Information
Additional Information:
This assay is in the 5'UTR region of the XM_021084778.1 transcripts.
Set Membership: |
![]() ![]() ![]() |
Panther Classification:
