Assay Details
Gene Symbol: PCBD2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_021084778.1 | XP_020940437.1 | 269 | 90 | ||
GenBank mRNA | AY610216.1 | - | 2 - 3 | 227 | 90 |

More Information
Set Membership: |
![]() ![]() ![]() |
Panther Classification:
