Assay Details
Gene Symbol: ICOSLG | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005657166.3 | XP_005657223.1 | 230 | 76 | ||
XM_005657167.3 | XP_005657224.1 | 230 | 76 |

More Information
Set Membership: |
![]() ![]() |
Panther Classification:
