Assay Details
Gene Symbol: ICOSLG | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005657167.3 | XP_005657224.1 | 1054 | 71 |

More Information
Set Membership: |
![]() ![]() |
Panther Classification:
