Assay Details
Gene Symbol: FGF10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_021077140.1 | XP_020932799.1 | 3385 | 72 |

More Information
Set Membership: |
![]() ![]() |
Panther Classification:
