Assay Details
Gene Symbol: SEMA7A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_021099054.1 | XP_020954713.1 | 235 | 92 |

More Information
Set Membership: |
![]() ![]() |
Panther Classification:
