Assay Details
Gene Symbol: SEMA7A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_021099054.1 | XP_020954713.1 | 388 | 107 | ||
XM_021099055.1 | XP_020954714.1 | 540 | 107 |

More Information
Additional Information:
This assay is in the 5'UTR region of the XM_021099055.1 transcripts.
Set Membership: |
![]() ![]() |
Panther Classification:
