Assay Details
Gene Symbol: PVR | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005653282.3 | XP_005653339.1 | 1065 | 95 | ||
XM_005653283.3 | XP_005653340.1 | 1067 | 95 | |||
XM_021094514.1 | XP_020950173.1 | 945 | 95 | |||
XM_021094515.1 | XP_020950174.1 | 942 | 95 |
