Assay Details
Gene Symbol: PVR | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005653282.3 | XP_005653339.1 | 1488 | 109 | ||
XM_005653283.3 | XP_005653340.1 | 1490 | 109 | |||
XM_021094514.1 | XP_020950173.1 | 1368 | 109 | |||
XM_021094515.1 | XP_020950174.1 | 1365 | 109 |

More Information
Set Membership: |
![]() ![]() |
