Assay Details
Gene Symbol: POLD4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005660637.3 | XP_005660694.3 | 344 | 89 | ||
GenBank mRNA | AK232254.1 | - | 1 - 2 | 291 | 89 | |
AK392779.1 | - | 1 - 2 | 244 | 89 | ||
AK397585.1 | - | 1 - 2 | 294 | 89 | ||
AY610249.1 | - | 1 - 2 | 233 | 89 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
Molecular Function -



