Assay Details
Gene Symbol: DMAC2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005655871.3 | XP_005655928.1 | 735 | 86 | ||
XM_005655872.3 | XP_005655929.1 | 735 | 86 | |||
GenBank mRNA | AK232262.1 | - | 1 - 2 | 47 | 86 | |
AK239042.1 | - | 1 - 2 | 47 | 86 | ||
AK345721.1 | - | 1 - 2 | 45 | 86 | ||
AK390071.1 | - | 1 - 2 | 49 | 86 | ||
AK395635.1 | - | 1 - 2 | 50 | 86 | ||
AK396329.1 | - | 1 - 2 | 48 | 86 | ||
AK396765.1 | - | 1 - 2 | 48 | 86 | ||
AK398186.1 | - | 1 - 2 | 47 | 86 |

More Information
Additional Information:
This assay is in the 5'UTR region of the XM_005655871.3, XM_005655872.3 transcripts.
Set Membership: |
![]() ![]() ![]() |
