Assay Details
Gene Symbol: MINDY2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_021094542.1 | XP_020950201.1 | 1761 | 98 | ||
XM_021094552.1 | XP_020950211.1 | 1665 | 98 | |||
GenBank mRNA | AK232627.1 | - | 7 - 8 | 1632 | 98 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
