Assay Details
Gene Symbol: NUDT8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003122437.4 | XP_003122485.1 | 1601 | 81 | ||
GenBank mRNA | AK232722.1 | - | 1 - 2 | 229 | 81 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
