Assay Details
Gene Symbol: NUDT8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003122437.4 | XP_003122485.1 | 1807 | 94 | ||
GenBank mRNA | AK232722.1 | - | 3 - 4 | 435 | 94 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Panther Classification:
