Assay Details
Gene Symbol: KLHL11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_021066993.1 | XP_020922652.1 | 856 | 118 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
