Assay Details
Gene Symbol: TRIB3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_013983169.2 | XP_013838623.2 | 294 | 113 |

More Information
Additional Information:
This assay is in the 5'UTR region of the XM_013983169.2 transcripts.
Set Membership: |
![]() ![]() |
Panther Classification:
