Assay Details
Gene Symbol: NENF | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_021063856.1 | XP_020919515.1 | 271 | 93 |

More Information
Set Membership: |
![]() ![]() ![]() |
Panther Classification:
