Assay Details
Gene Symbol: SCN4B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003129902.4 | XP_003129950.1 | 763 | 81 | ||
XM_013977487.2 | XP_013832941.1 | 859 | 81 |

More Information
Set Membership: |
![]() ![]() ![]() |
Panther Classification:
