Assay Details
Gene Symbol: KCNC3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_021094790.1 | XP_020950449.1 | 2190 | 97 |

More Information
Set Membership: |
![]() ![]() |
Panther Classification:
