Assay Details
Gene Symbol: KCNC3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005664780.3 | XP_005664837.1 | 2249 | 100 | ||
XM_021094790.1 | XP_020950449.1 | 2375 | 100 |

More Information
Set Membership: |
![]() ![]() ![]() |
Panther Classification:
