Assay Details
Gene Symbol: MTSS2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_021093809.1 | XP_020949468.1 | 1408 | 73 | ||
XM_021093810.1 | XP_020949469.1 | 1399 | 73 | |||
XM_021093811.1 | XP_020949470.1 | 1319 | 73 | |||
XM_021093812.1 | XP_020949471.1 | 1303 | 73 |
