Assay Details
Gene Symbol: MTSS2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_021093811.1 | XP_020949470.1 | 941 | 87 | ||
XM_021093812.1 | XP_020949471.1 | 925 | 87 |

UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: MTSS2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_021093811.1 | XP_020949470.1 | 941 | 87 | ||
XM_021093812.1 | XP_020949471.1 | 925 | 87 |