Assay Details
Gene Symbol: ZHX1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001928633.6 | XP_001928668.1 | 424 | 100 | ||
XM_013996503.2 | XP_013851957.1 | 417 | 100 | |||
XM_021088958.1 | XP_020944617.1 | 414 | 100 |

More Information
Additional Information:
This assay is in the 5'UTR region of the XM_001928633.6, XM_013996503.2, XM_021088958.1 transcripts.
Set Membership: |
![]() ![]() ![]() |
Panther Classification:
