Assay Details
Gene Symbol: NOXO1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_013987814.2 | XP_013843268.1 | 639 | 101 | ||
XM_013987815.2 | XP_013843269.1 | 636 | 101 | |||
XM_013987816.2 | XP_013843270.1 | 639 | 101 |

More Information
Set Membership: |
![]() ![]() |
