Assay Details
Gene Symbol: NOXO1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_013987814.2 | XP_013843268.1 | 822 | 82 | ||
XM_013987815.2 | XP_013843269.1 | 819 | 82 |

UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: NOXO1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_013987814.2 | XP_013843268.1 | 822 | 82 | ||
XM_013987815.2 | XP_013843269.1 | 819 | 82 |