Assay Details
Gene Symbol: LOC100154044 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001928880.6 | XP_001928915.1 | 647 | 85 | ||
XM_005666256.3 | XP_005666313.1 | 633 | 85 | |||
XM_013978205.2 | XP_013833659.1 | 648 | 85 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
