Assay Details
Gene Symbol: MYCL | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003127827.5 | XP_003127875.2 | 193 | 65 |

More Information
Additional Information:
This assay is in the 5'UTR region of the XM_003127827.5 transcripts.
Set Membership: |
![]() ![]() |
Panther Classification:
