Assay Details
Gene Symbol: MYCL | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003127827.5 | XP_003127875.2 | 695 | 103 | ||
XM_021095995.1 | XP_020951654.1 | 843 | 103 | |||
GenBank mRNA | HQ845207.1 | - | 1 - 2 | 492 | 103 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Panther Classification:
