Assay Details
Gene Symbol: MOB2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_021082597.1 | XP_020938256.1 | 96 | 52 |

More Information
Additional Information:
This assay is in the 5'UTR region of the XM_021082597.1 transcripts.
Set Membership: |
![]() ![]() |
Panther Classification:
