Assay Details
Gene Symbol: MOB2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_021082596.1 | XP_020938255.1 | 508 | 66 | ||
XM_021082597.1 | XP_020938256.1 | 485 | 66 |

More Information
Set Membership: |
![]() ![]() ![]() |
Panther Classification:
