Assay Details
Gene Symbol: HECA | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_021087304.1 | XP_020942963.1 | 1596 | 83 |

UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: HECA | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_021087304.1 | XP_020942963.1 | 1596 | 83 |